Anti-B4GALT2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-762
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:B4GALT2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60909
- Antigen Synonyms:4-galactosyltransferase 2|4-galactosyltransferase|4-GalTase 2|B4Gal T2|b4Gal-T2|B4GT2_HUMAN|Beta 1,4 galactosyltransferase 2|Beta 1,4 GalTase 2|Beta 4 GalT2|Beta N acetylglucosaminyl glycolipid beta 1,4 galactosyltransferase 2|Beta-1|Beta-N-acetylglucosaminyl-glycolipid beta-1|Beta4Gal T2|Beta4Gal-T2|Nal synthase|UDP Gal:beta GlcNAc beta 1,4 galactosyltransferase 2|UDP Gal:betaGlcNAc beta 1,4 galactosyltransferase polypeptide 2|UDP galactose:beta N acetylglucosamine beta 1,4 galactosyltransferase 2|UDP-Gal:beta-GlcNAc beta-1|UDP-galactose:beta-N-acetylglucosamine beta-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-1,4-galactosyltransferase 2 protein (271-372AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: B4GALT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human