Anti-CRTAC1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77853-590
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRTAC1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q9NQ79
- Antigen Synonyms:68 kDa chondrocyte expressed protein|68 kDa chondrocyte-expressed protein|Acidic secreted protein in cartilage|ASPIC|ASPIC1|Cartilage acidic protein 1|Cartilage acidic protein 1 precursor|CEP 68|CEP-68|CEP68|Chondrocyte expressed protein 68 kDa CEP 68|CRAC1_HUMAN|Protein CRTAC1-B|W307
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cartilage acidic protein 1 protein (309-661AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CRTAC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human