Anti-ADH1B Rabbit Recombinant Polyclonal Antibody
77850-775
New Product
- Antigen Symbol:ADH1B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P00325
- Antigen Synonyms:ADH beta subunit|ADH1B|ADH1B_HUMAN|ADH2|Alcohol dehydrogenase 1B|alcohol dehydrogenase 2 (class I)|beta polypeptide|Alcohol dehydrogenase 2|Alcohol dehydrogenase subunit beta|Aldehyde reductase|DKFZp686C06125|OTTHUMP00000220192
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alcohol dehydrogenase 1B protein (2-375AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ADH1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human