Anti-PTBP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-738
- 77844-737
- Antigen Symbol:
- PTBP2
- PTBP2
- Antigen Synonyms:
- brPTB|FLJ34897|MIBP|Neural polypyrimidine tract binding protein|Neural polypyrimidine tract-binding protein|Neurally enriched homolog of PTB|Neurally-enriched homolog of PTB|nPTB 5|nPTB 6|nPTB 7|nPTB 8|nPTB|nPTB5|nPTB6|nPTB7|nPTB8|Polypyrimidine tract binding protein 2|Polypyrimidine tract-binding protein 2|PTB|PTB like|PTB like protein|PTB-like protein|PTBLP|PTBP 2|PTBP2_HUMAN|Splicing regulator
- brPTB|FLJ34897|MIBP|Neural polypyrimidine tract binding protein|Neural polypyrimidine tract-binding protein|Neurally enriched homolog of PTB|Neurally-enriched homolog of PTB|nPTB 5|nPTB 6|nPTB 7|nPTB 8|nPTB|nPTB5|nPTB6|nPTB7|nPTB8|Polypyrimidine tract binding protein 2|Polypyrimidine tract-binding protein 2|PTB|PTB like|PTB like protein|PTB-like protein|PTBLP|PTBP 2|PTBP2_HUMAN|Splicing regulator
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKA9
- Q9UKA9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Polypyrimidine tract-binding protein 2 protein (13-289AA)
- Recombinant Human Polypyrimidine tract-binding protein 2 protein (13-289AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice