Anti-Adh5 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77850-785
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Adh5
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:P28474
- Antigen Synonyms:Adh-2|Adh2|Alcohol dehydrogenase class-3|EC 1.1.1.1|Alcohol dehydrogenase 2|Alcohol dehydrogenase 5|Alcohol dehydrogenase B2|ADH-B2|Alcohol dehydrogenase class-III|Glutathione-dependent formaldehyde dehydrogenase|FALDH|FDH|GSH-FDH|EC 1.1.1.-|S-(hydroxymethyl)glutathione dehydrogenase|EC 1.1.1.284
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Mouse Alcohol dehydrogenase class-3 protein (2-374AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: Adh5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse