Anti-ATP5F1A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-594
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5F1A
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P25705
- Antigen Synonyms:ATP synthase alpha chain|mitochondrial|ATP synthase subunit alpha|ATP synthase subunit alpha mitochondrial|ATP synthase|H+ transporting|mitochondrial F1 complex|alpha subunit 1|cardiac muscle|alpha subunit|1|isoform 1|isoform 2|non-cardiac muscle-like 2|ATP sythase (F1 ATPase) alpha subunit|ATP5A|Atp5a1|ATP5AL2|ATPA_HUMAN|ATPM|Epididymis secretory sperm binding protein Li 123m|hATP1|HEL-S-123m|MC5DN4|Mitochondrial ATP synthetase|Mitochondrial ATP synthetase oligomycin resistant|Modifier of Min 2|Modifier of Min 2 mouse homolog|mouse|homolog of|MOM2|OMR|ORM|OTTHUMP00000163475
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP synthase subunit alpha, mitochondrial protein (59-257AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATP5F1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human