Anti-DAP3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-974
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DAP3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P51398
- Antigen Synonyms:28S ribosomal protein S29|28S ribosomal protein S29 mitochondrial|bMRP10|DAP 3|DAP-3|Death associated protein 3|Death-associated protein 3|DKFZp686G12159|FLJ12817|Ionizing radiation resistance conferring protein|MGC126058|MGC126059|Mitochondrial 28S ribosomal protein S29|mitochondrial|MITOCHONDRIAL RIBOSOMAL PROTEIN S29|MRP S29|MRP-S29|MRPS29|OTTHUMP00000033480|OTTHUMP00000033481|PSD-95/SAP90-binding protein 3|RT29_HUMAN|S29mt|SAP90/PSD-95-associated protein 3|SAPAP3
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human 28S ribosomal protein S29, mitochondrial protein (159-398AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: DAP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human