Anti-CRADD Rabbit Recombinant polyclonal Antibody (Biotinylated)
Catalog # 77853-527
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CRADD
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P78560
- Antigen Synonyms:CASP2 and RIPK1 domain containing adaptor with death domain |Caspase and RIP adapter with death domain|Caspase and RIP adaptor with death domain |CRADD_HUMAN|Death adaptor molecule RAIDD |Death domain containing protein CRADD |Death domain-containing protein MGC9163|RIP associated ICH1/CED3 homologous protein with death domain |RIP associated protein with a death domain |RIP-associated protein with a death domain
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Death domain-containing protein CRADD protein (1-199AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: CRADD
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human