Anti-ELOVL2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ELOVL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-576
- 77843-575
- Antigen Symbol:
- ELOVL2
- ELOVL2
- Antigen Synonyms:
- 3 keto acyl CoA synthase 3-keto acyl-CoA synthase Elongation of very long chain fatty acids (FEN1/Elo2|SUR4/Elo3|yeast) like 2|Elongation of very long chain fatty acids protein 2|ELOV2_HUMAN|ELOVL fatty acid elongase 2|FLJ20334|OTTHUMP00000016029|SSC2
- 3 keto acyl CoA synthase 3-keto acyl-CoA synthase Elongation of very long chain fatty acids (FEN1/Elo2|SUR4/Elo3|yeast) like 2|Elongation of very long chain fatty acids protein 2|ELOV2_HUMAN|ELOVL fatty acid elongase 2|FLJ20334|OTTHUMP00000016029|SSC2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NXB9
- Q9NXB9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human elongation of very long chain fatty acids protein 2 protein (91-174AA)
- Recombinant human elongation of very long chain fatty acids protein 2 protein (91-174AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice