Anti-MTCH2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MTCH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-654
- 77846-653
- Antigen Symbol:
- MTCH2
- MTCH2
- Antigen Synonyms:
- 2310034D24Rik|HSPC032|Met-induced mitochondrial protein|MIMP|Mitochondrial carrier 2|Mitochondrial carrier homolog 2|MTCH2_HUMAN|OTTHUMP00000234441|OTTHUMP00000234442
- 2310034D24Rik|HSPC032|Met-induced mitochondrial protein|MIMP|Mitochondrial carrier 2|Mitochondrial carrier homolog 2|MTCH2_HUMAN|OTTHUMP00000234441|OTTHUMP00000234442
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6C9
- Q9Y6C9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitochondrial carrier homolog 2 protein (31-131AA)
- Recombinant Human Mitochondrial carrier homolog 2 protein (31-131AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice