Bovine Hyaluronidase (from Testes)
Catalog # 82603-950
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:37326-33-3
- Source:Testes
- Species:Bovine
- Size:25 kU
- Storage Conditions:OK to freeze
- Enzyme Name:Hyaluronidase
- Enzyme Synonyms:Hyaluronidase