Human Chymotrypsin (from Pancreas)
Catalog # 80051-044
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9004-07-3
- Source:Pancreas
- Species:Human
- Size:50 μg
- Storage Conditions:Safe to freeze
- Enzyme Name:Chymotrypsin
- Purity:≥95% (SDS-PAGE)