Anti-TAB1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: TAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77848-783
- 77848-782
- Antigen Symbol:
- TAB1
- TAB1
- Antigen Synonyms:
- 2310012M03Rik|3'-MAP3K7IP 1|MAP3K7IP1|MGC57664|Mitogen activated protein kinase kinase kinase 7 interacting protein 1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TAB 1|TAB1_HUMAN|TAK1 binding protein 1|TAK1-binding protein 1|TGF beta activated kinase 1 binding protein 1|TGF-beta activated kinase 1/MAP3K7 binding protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|TGF-beta-activated kinase 1-binding protein 1|Transforming growth factor beta activated kinase binding protein 1
- 2310012M03Rik|3'-MAP3K7IP 1|MAP3K7IP1|MGC57664|Mitogen activated protein kinase kinase kinase 7 interacting protein 1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TAB 1|TAB1_HUMAN|TAK1 binding protein 1|TAK1-binding protein 1|TGF beta activated kinase 1 binding protein 1|TGF-beta activated kinase 1/MAP3K7 binding protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|TGF-beta-activated kinase 1-binding protein 1|Transforming growth factor beta activated kinase binding protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15750
- Q15750
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TAB1
- Fusion protein of Human TAB1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice