Anti-CD59 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-705
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD59
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P13987
- Antigen Synonyms:16.3A5|1F5|1F5 antigen|20 kDa homologous restriction factor|CD 59|CD_antigen=CD59 antigen|CD59 antigen complement regulatory protein|CD59 antigen p18 20|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5|EJ16|EJ30|EL32 and G344)|CD59 glycoprotein|CD59 molecule|CD59 molecule complement regulatory protein|CD59_HUMAN|Cd59a|Complement regulatory protein|EL32|FLJ38134|FLJ92039|G344|HRF 20|HRF-20|HRF20|Human leukocyte antigen MIC11|Ly 6 like protein|Lymphocytic antigen CD59/MEM43|MAC inhibitory protein|MAC IP|MAC-inhibitory protein|MAC-IP|MACIF|MACIP|MEM43|MEM43 antigen|Membrane attack complex (MAC) inhibition factor|Membrane attack complex inhibition factor|Membrane inhibitor of reactive lysis|MGC2354|MIC11|MIN1|MIN2|MIN3|MIRL|MSK21|p18 20|Protectin|Surface antigen recognized by monoclonal 16.3A5|T cell activating protein
- Storage Buffer:0.1 mol/L NaCl, 15 mmol/L NaN₃, pH 7.2
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CD59 glycoprotein protein (26-102AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CD59
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse