Anti-DMBT1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:10 to 1:50
Type:
Antigen: DMBT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-403
- 77847-404
- Antigen Symbol:
- DMBT1
- DMBT1
- Antigen Synonyms:
- Apactin|CRP|CRP-[a]|CRP-[b]|CRP-ductin|Crpd|DBMT1|Deleted in malignant brain tumors 1|Deleted in malignant brain tumors 1 protein|DMBT 1|DMBT1 prototype precursor|DMBT1/6kb.1 protein precursor|DMBT1/8kb.1 protein|DMBT1/8kb.2 protein precursor|DMBT1_HUMAN|Ebnerin|FLJ61058|Glycoprotein 300|Glycoprotein 340|Gp 340|Gp-340|gp300|GP340|Gp340 variant protein|Hensin|MGC164738|Mucin-like glycoprotein|Muclin|OTTHUMP00000020654|OTTHUMP00000020655|OTTHUMP00000020656|OTTHUMP00000020657|OTTHUMP00000020658|OTTHUMP00000020659|OTTHUMP00000020660|OTTHUMP00000020661|OTTHUMP00000020662|OTTHUMP00000020663|p80|Pancrin|SAG|Salivary agglutinin|Surfactant pulmonary associated D binding protein|Surfactant pulmonary associated protein D binding protein|Surfactant pulmonary-associated D-binding protein|Vomeroglandin
- Apactin|CRP|CRP-[a]|CRP-[b]|CRP-ductin|Crpd|DBMT1|Deleted in malignant brain tumors 1|Deleted in malignant brain tumors 1 protein|DMBT 1|DMBT1 prototype precursor|DMBT1/6kb.1 protein precursor|DMBT1/8kb.1 protein|DMBT1/8kb.2 protein precursor|DMBT1_HUMAN|Ebnerin|FLJ61058|Glycoprotein 300|Glycoprotein 340|Gp 340|Gp-340|gp300|GP340|Gp340 variant protein|Hensin|MGC164738|Mucin-like glycoprotein|Muclin|OTTHUMP00000020654|OTTHUMP00000020655|OTTHUMP00000020656|OTTHUMP00000020657|OTTHUMP00000020658|OTTHUMP00000020659|OTTHUMP00000020660|OTTHUMP00000020661|OTTHUMP00000020662|OTTHUMP00000020663|p80|Pancrin|SAG|Salivary agglutinin|Surfactant pulmonary associated D binding protein|Surfactant pulmonary associated protein D binding protein|Surfactant pulmonary-associated D-binding protein|Vomeroglandin
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UGM3
- Q9UGM3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human DMBT1
- Synthetic peptide of Human DMBT1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice