Anti-ACOT11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: ACOT11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-079
- 77847-078
- Antigen Symbol:
- ACOT11
- ACOT11
- Antigen Synonyms:
- ACO11_HUMAN| ACOT 11| Acyl CoA thioester hydrolase 11| Acyl CoA thioesterase 11| Acyl coenzyme A thioesterase 11| Acyl-CoA thioester hydrolase 11| Acyl-CoA thioesterase 11| Acyl-coenzyme A thioesterase 11| Adipose associated thioesterase| Adipose-associated thioesterase| BFIT 1| BFIT 2| BFIT| BFIT1| BFIT2| Brown fat inducible thioesterase| Brown fat-inducible thioesterase| KIAA0707| OTTHUMP00000046722| StAR related lipid transfer (START) domain containing 14| STARD 14| STARD14| START domain containing 14| THEA| THEM 1| THEM1| Thioesterase adipose associated| Thioesterase superfamily member 1
- ACO11_HUMAN| ACOT 11| Acyl CoA thioester hydrolase 11| Acyl CoA thioesterase 11| Acyl coenzyme A thioesterase 11| Acyl-CoA thioester hydrolase 11| Acyl-CoA thioesterase 11| Acyl-coenzyme A thioesterase 11| Adipose associated thioesterase| Adipose-associated thioesterase| BFIT 1| BFIT 2| BFIT| BFIT1| BFIT2| Brown fat inducible thioesterase| Brown fat-inducible thioesterase| KIAA0707| OTTHUMP00000046722| StAR related lipid transfer (START) domain containing 14| STARD 14| STARD14| START domain containing 14| THEA| THEM 1| THEM1| Thioesterase adipose associated| Thioesterase superfamily member 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WXI4
- Q8WXI4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ACOT11
- Fusion protein of Human ACOT11
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice