Anti-NAA10 Rabbit Recombinant Polyclonal Antibody
77851-329
New Product
- Antigen Symbol:NAA10
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P41227
- Antigen Synonyms:Alpha N acetyltransferase 1A|ARD1|ARD1 homolog N acetyltransferase (S. cerevisiae)|ARD1 homolog A N acetyltransferase (S. cerevisiae)|ARD1 homolog A N acetyltransferase|ARD1A|DXS707|MGC71248|N acetyltransferase ARD1, human homolog of|N alpha acetyltransferase 10 NatA catalytic subunit|N terminal acetyltransferase complex ARD1 subunit homolog A|N(alpha) acetyltransferase 10 NatA catalytic subunit|N-alpha-acetyltransferase 10|N-terminal acetyltransferase complex ARD1 subunit homolog A|Naa10|NAA10_HUMAN|NatA catalytic subunit|TE2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human N-alpha-acetyltransferase 10 protein (161-235AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: NAA10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human