Anti-AKR1C1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-972
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AKR1C1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:Q04828
- Antigen Synonyms:2-dihydrobenzene-1|2-diol dehydrogenase|20 alpha (3 alpha) hydroxysteroid dehydrogenase|20 ALPHA HSD|20 alpha hydroxysteroid dehydrogenase|20-alpha-HSD|20-alpha-hydroxysteroid dehydrogenase|20ALPHAHSD|2ALPHAHSD|AK1C1|AK1C1_HUMAN|AKR1C1|Aldo keto reductase family 1 member C1|aldo-keto reductase C|Aldo-keto reductase family 1 member C1|Aldo-keto reductase family|1 member 1|C9|Chlordecone reductase homolog|Chlordecone reductase homolog HAKRC|DD1|DD1/DD2|DDH|DDH1|Dihydrodiol dehydrogenase 1|Dihydrodiol dehydrogenase 1/2|dihydrodiol dehydrogenase 1|20-alpha (3-alpha)-hydroxysteroid dehydrogenase|dihydrodiol dehydrogenase isoform DD1|Dihydrodiol dehydrogenase|type 1|H37|HAKRC|HBAB|Hepatic dihydrodiol dehydrogenase|High affinity hepatic bile acid-binding protein|High-affinity hepatic bile acid-binding protein|Indanol dehydrogenase|MBAB|MGC8954|Trans-1|Trans-1,2 dihydrobenzene 1,2 diol dehydrogenase|Type II 3 alpha hydroxysteroid dehydrogenase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aldo-keto reductase family 1 member C1 protein (219-323AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF:1:50 to 1:200
Type:
Antigen: AKR1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human