Anti-ATG3 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-532
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATG3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NT62
- Antigen Synonyms:2610016C12Rik|Apg 3|APG3|APG3 autophagy 3 like|APG3 like|APG3, S. cerevisiae, homolog of|APG3-like|APG3L|Apg3p|ATG 3|ATG3 autophagy related 3 homolog|ATG3 autophagy related 3 homolog (S. cerevisiae)|ATG3_HUMAN|Autophagy 3, S. cerevisiae, homolog of|Autophagy Apg3p/Aut1p like|autophagy related 3|Autophagy related protein 3|Autophagy-related protein 3|DKFZp564M1178|FLJ22125|hApg3|MGC15201|OTTHUMP00000214547|OTTHUMP00000214548|PC3 96|Protein PC3-96|Ubiquitin-like-conjugating enzyme ATG3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Ubiquitin-like-conjugating enzyme ATG3 protein (1-314AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: ATG3
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human