Anti-HOXD10 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-599
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXD10
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P28358
- Antigen Synonyms:AI385591|AI874987|Homeo box 4D|Homeo box D10|Homeobox D10|Homeobox protein Hox-4D|Homeobox protein Hox-4E|Homeobox protein Hox-D10|Hox 4.4|Hox 4.5|Hox 4D|Hox 4E|Hox 5.3|HOX4D|HOX4E|HOXD10|HXD10|HXD10_HUMAN|OTTMUSP00000017845|RP23-313J15.9
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Homeobox protein Hox-D10 protein (1-240AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: HOXD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse