Anti-NACA2 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NACA2
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-050
- 77843-049
- Antigen Symbol:
- NACA2
- NACA2
- Antigen Synonyms:
- NACAL|Nascent polypeptide-associated complex subunit alpha-2|Alpha-NAC-like|Hom s 2.01|Nascent polypeptide-associated complex subunit alpha-like|NAC-alpha-like
- NACAL|Nascent polypeptide-associated complex subunit alpha-2|Alpha-NAC-like|Hom s 2.01|Nascent polypeptide-associated complex subunit alpha-like|NAC-alpha-like
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H009
- Q9H009
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Nascent polypeptide-associated complex subunit alpha-2 protein (1-215AA)
- Recombinant Human Nascent polypeptide-associated complex subunit alpha-2 protein (1-215AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice