Anti-PACSIN3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: PACSIN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-114
- 77845-115
- Antigen Symbol:
- PACSIN3
- PACSIN3
- Antigen Synonyms:
- Endophilin 1|Endophilin I|FLJ17861|OTTHUMP00000234262|OTTHUMP00000234263|OTTHUMP00000234264|PACN3_HUMAN|Protein kinase C and casein kinase substrate in neurons 3|Protein kinase C and casein kinase substrate in neurons protein 3|SDPIII|SH3 domain containing protein 6511|SH3 domain-containing protein 6511|syndapin III
- Endophilin 1|Endophilin I|FLJ17861|OTTHUMP00000234262|OTTHUMP00000234263|OTTHUMP00000234264|PACN3_HUMAN|Protein kinase C and casein kinase substrate in neurons 3|Protein kinase C and casein kinase substrate in neurons protein 3|SDPIII|SH3 domain containing protein 6511|SH3 domain-containing protein 6511|syndapin III
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKS6
- Q9UKS6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein kinase C and casein kinase substrate in neurons protein 3 protein (1-300AA)
- Recombinant Human Protein kinase C and casein kinase substrate in neurons protein 3 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice