Anti-DCLK3 Rabbit Polyclonal Antibody
Catalog # 77849-813
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DCLK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:liquid
- Gene ID:Q9C098
- Antigen Synonyms:BC056929|C730036H08|CLICK I and II related|CLr|Click I II related|CLR|Dcamkl3|DCDC3C|DCK3|Dclk3|DCLK3_HUMAN|Doublecortin and CaM kinase like 3|Doublecortin domain-containing protein 3C|Doublecortin-like and CAM kinase-like 3|Doublecortin-like kinase 3|hCG_16695|KIAA1765|RGD1309232|Serine/threonine-protein kinase DCLK3
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human DCLK3.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: DCLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human