Anti-CSGALNACT2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77853-657
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CSGALNACT2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q8N6G5
- Antigen Synonyms:4-N-acetylgalactosaminyltransferase 2|Beta4GalNAcT-2|CGAT2_HUMAN|CHGN2|Chondroitin beta-1|Chondroitin sulfate N-acetylgalactosaminyltransferase 2|Csgalnact2|DKFZp686H13226|FLJ43310|GALNACT 2|GalNAcT-2|GALNACT2|MGC40204|PRO0082
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chondroitin sulfate N-acetylgalactosaminyltransferase 2 protein (33-270AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CSGALNACT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human