Anti-NLE1 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: NLE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-554
- 77843-553
- Antigen Symbol:
- NLE1
- NLE1
- Antigen Synonyms:
- AL022765|BC018399|FLJ10458|NLE 1|Nle|NLE1_HUMAN|Notchless gene homolog|Notchless homolog 1 (Drosophila)|Notchless homolog 1|Notchless protein homolog 1
- AL022765|BC018399|FLJ10458|NLE 1|Nle|NLE1_HUMAN|Notchless gene homolog|Notchless homolog 1 (Drosophila)|Notchless homolog 1|Notchless protein homolog 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVX2
- Q9NVX2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Notchless protein homolog 1 protein (2-485AA)
- Recombinant Human Notchless protein homolog 1 protein (2-485AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice