Anti-BRCC3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-943
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BRCC3
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P46736
- Antigen Synonyms:BRCA1-A complex subunit BRCC36|BRCA1/BRCA2 containing complex subunit 3|BRCA1/BRCA2 containing complex subunit 36|BRCA1/BRCA2-containing complex subunit 3|BRCA1/BRCA2-containing complex subunit 36|BRCC3_HUMAN|BRCC36|BRISC complex subunit BRCC36|C6.1A|Chromosome X open reading frame 53|CXorf53|Lys-63-specific deubiquitinase BRCC36|OTTHUMP00000061450
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Lys-63-specific deubiquitinase BRCC36 protein (14-316AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: BRCC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human