Anti-EIF3J Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-610
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF3J
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75822
- Antigen Synonyms:eIF 3 alpha|eIF-3-alpha|eIF3 alpha|eIF3 p35|EIF3J_HUMAN|EIF3S 1|EIF3S1|Eukaryotic translation initiation factor 3 subunit 1|Eukaryotic translation initiation factor 3 subunit J|Eukaryotic translation initiation factor 3|subunit 1 (alpha|35kD)|subunit 1 alpha|35kD
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Eukaryotic translation initiation factor 3 subunit J protein (5-115AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EIF3J
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human