Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ASIP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-ASIP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-478
Supplier:  AFG Bioscience
New Product
undefined
Anti-ASIP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-478
Supplier:  AFG Bioscience
Supplier Number:  A13295-50UG
New Product

Specifications

  • Antigen Symbol:
    ASIP
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    P42127
  • Antigen Synonyms:
    AGTI|AGTIL|ASP|Agouti-signaling protein|Agouti switch protein
  • Storage Buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Agouti-signaling protein (23-112AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: ASIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human