Anti-ATP5B Rabbit Recombinant Polyclonal Antibody
77851-601
New Product
- Antigen Symbol:ATP5B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P06576
- Antigen Synonyms:ATP 5B|ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide|ATP synthase subunit beta mitochondrial|ATP synthase subunit beta|mitochondrial|ATPB|ATPB_HUMAN|ATPMB|ATPSB|Epididymis secretory protein Li 271|HEL-S-271|Mitochondrial ATP synthase beta subunit|Mitochondrial ATP Synthase Subunit Beta|Mitochondrial ATP synthetase beta subunit
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human ATP synthase subunit beta, mitochondrial protein (230-529AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ATP5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat