Anti-OAS3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-673
- 77846-674
- Antigen Symbol:
- OAS3
- OAS3
- Antigen Synonyms:
- (2-5'')oligo(A) synthase 3|2 5' oligo(A) synthetase 3|2 5'LIGO|2 5A synthetase 3|2''-5''-oligoadenylate synthase 3|2-5A synthase 3|MGC133260|OAS3 2' 5' oligoadenylate synthetase 3, 100kDa|OAS3_HUMAN|p100|p100 OAS|p100OAS
- (2-5'')oligo(A) synthase 3|2 5' oligo(A) synthetase 3|2 5'LIGO|2 5A synthetase 3|2''-5''-oligoadenylate synthase 3|2-5A synthase 3|MGC133260|OAS3 2' 5' oligoadenylate synthetase 3, 100kDa|OAS3_HUMAN|p100|p100 OAS|p100OAS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6K5
- Q9Y6K5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 2\'-5\'-oligoadenylate synthase 3 protein (424-565AA)
- Recombinant Human 2\'-5\'-oligoadenylate synthase 3 protein (424-565AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice