Anti-CASP2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77852-386
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KLC4
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NSK0
- Antigen Synonyms:Kinesin light chain 4|kinesin like 8|Kinesin like protein 8|Kinesin-like protein 8|KLC 4|KLC4|KLC4_HUMAN|KnsI8|KNSL8
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Kinesin light chain 4 protein (401-619AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human