Anti-CSGALNACT2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: CSGALNACT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-961
- 77849-962
- Antigen Symbol:
- CSGALNACT2
- CSGALNACT2
- Antigen Synonyms:
- 4-N-acetylgalactosaminyltransferase 2|Beta4GalNAcT-2|CGAT2_HUMAN|CHGN2|Chondroitin beta-1|Chondroitin sulfate N-acetylgalactosaminyltransferase 2|DKFZp686H13226|FLJ43310|GALNACT 2|GalNAcT-2|GALNACT2|MGC40204|PRO0082
- 4-N-acetylgalactosaminyltransferase 2|Beta4GalNAcT-2|CGAT2_HUMAN|CHGN2|Chondroitin beta-1|Chondroitin sulfate N-acetylgalactosaminyltransferase 2|DKFZp686H13226|FLJ43310|GALNACT 2|GalNAcT-2|GALNACT2|MGC40204|PRO0082
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N6G5
- Q8N6G5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CSGALNACT2
- Fusion protein of Human CSGALNACT2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice