Anti-NEUROG3 Rabbit Polyclonal Antibody
Catalog # 77848-730
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NEUROG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9Y4Z2
- Antigen Synonyms:ATH4B|Atoh5|bHLHa7|Class A basic helix-loop-helix protein 7|Math4B|NEUROG3|Neurogenin-3|Neurogenin3|NGN-3|ngn3|NGN3_HUMAN|OTTHUMP00000019730|Protein atonal homolog 5
- Storage Buffer:PBS, 0.05% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Full length fusion protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:3000, IHC:1:50 to 1:100
Type:
Antigen: NEUROG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat