Anti-SRM Rabbit Recombinant polyclonal Antibody
Catalog # 77857-952
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SRM
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P19623
- Antigen Synonyms:OTTHUMP00000002170|PAPT|Putrescine aminopropyltransferase|SPDSY|SPEE_HUMAN|Spermidine synthase 1|Spermidine synthase|SPS1|SRML1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Spermidine synthase protein (17-302AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: SRM
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human