Anti-SRPK3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: SRPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-307
- 77847-306
- Antigen Symbol:
- SRPK3
- SRPK3
- Antigen Synonyms:
- MSSK-1|MSSK1|Muscle specific serine kinase 1|Muscle-specific serine kinase 1|Serine/arginine rich protein specific kinase 3|Serine/arginine-rich protein-specific kinase 3|Serine/threonine protein kinase 23|Serine/threonine protein kinase Serine/threonine-protein kinase 23|SFRS protein kinase 3|SR-protein-specific kinase 3|SRPK3_HUMAN|SRSF protein kinase 3|STK 23|STK23
- MSSK-1|MSSK1|Muscle specific serine kinase 1|Muscle-specific serine kinase 1|Serine/arginine rich protein specific kinase 3|Serine/arginine-rich protein-specific kinase 3|Serine/threonine protein kinase 23|Serine/threonine protein kinase Serine/threonine-protein kinase 23|SFRS protein kinase 3|SR-protein-specific kinase 3|SRPK3_HUMAN|SRSF protein kinase 3|STK 23|STK23
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPE1
- Q9UPE1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SRPK3
- Fusion protein of Human SRPK3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice