Anti-AGTR1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77850-894
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AGTR1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P30556
- Antigen Synonyms:AG2S|Agtr 1|Agtr1|AGTR1_HUMAN|Agtr1a|AGTR1B|Ang II|Angiotensin II receptor type 1|Angiotensin II type-1 receptor|Angiotensin receptor 1|Angiotensin receptor 1B|AT 1B|AT 1r|AT1|At1a|AT1AR|AT1B|AT1BR|AT1R|AT2R1|AT2R1A|AT2R1B|HAT1R|Type 1 angiotensin II receptor|Type 1B angiotensin II receptor|Type-1 angiotensin II receptor
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Type-1 angiotensin II receptor protein (306-356AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AGTR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human