Anti-LCP1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77858-864
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LCP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P13796
- Antigen Synonyms:CP64|L plastin|L-plastin|Larval cuticle protein 1|LC64P|LCP-1|LPL|Lplastin|Lymphocyte cytosolic protein 1|Plastin 2|Plastin-2|PLS2|PLSL_HUMAN
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Plastin-2 protein (2-627AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500, IP: 1:200 to 1:2000
Type:
Antigen: LCP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human