Anti-BCL6 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77847-076
- 77847-077
- Antigen Symbol:
- BCL6
- BCL6
- Antigen Synonyms:
- B cell CLL/lymphoma 6|B cell lymphoma 6 protein|B-cell lymphoma 5 protein|B-cell lymphoma 6 protein|BCL 5|Bcl 6|BCL-5|BCL-6|BCL5|BCL6|BCL6_HUMAN|BCL6A|cys his2 zinc finger transcription factor|cys-his2 zinc finger transcription factor|LAZ 3|LAZ 3 protein|LAZ3|Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3)|Lymphoma associated zinc finger gene on chromosome 3|Protein LAZ-3|ZBTB 27|ZBTB27|Zinc finger and BTB domain containing protein 27|Zinc finger and BTB domain-containing protein 27 (ZBTB27)|Zinc finger and BTB domain-containing protein 27|Zinc Finger Protein 51 (ZNF51)|Zinc finger protein 51|zinc finger transcription factor BCL6S|ZNF 51|ZNF51
- B cell CLL/lymphoma 6|B cell lymphoma 6 protein|B-cell lymphoma 5 protein|B-cell lymphoma 6 protein|BCL 5|Bcl 6|BCL-5|BCL-6|BCL5|BCL6|BCL6_HUMAN|BCL6A|cys his2 zinc finger transcription factor|cys-his2 zinc finger transcription factor|LAZ 3|LAZ 3 protein|LAZ3|Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3)|Lymphoma associated zinc finger gene on chromosome 3|Protein LAZ-3|ZBTB 27|ZBTB27|Zinc finger and BTB domain containing protein 27|Zinc finger and BTB domain-containing protein 27 (ZBTB27)|Zinc finger and BTB domain-containing protein 27|Zinc Finger Protein 51 (ZNF51)|Zinc finger protein 51|zinc finger transcription factor BCL6S|ZNF 51|ZNF51
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41182
- P41182
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human BCL6
- Synthetic peptide of Human BCL6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice