Anti-MAGEB18 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: MAGEB18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77849-158
- 77849-157
- Antigen Symbol:
- MAGEB18
- MAGEB18
- Antigen Synonyms:
- B430216B18Rik|MAGBI_HUMAN|MAGE-B18 antigen|MAGEB18 antigen|Melanoma antigen family B|18|Melanoma-associated antigen B18|MGC114496|RGD1560369|RP23-289C5.3
- B430216B18Rik|MAGBI_HUMAN|MAGE-B18 antigen|MAGEB18 antigen|Melanoma antigen family B|18|Melanoma-associated antigen B18|MGC114496|RGD1560369|RP23-289C5.3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96M61
- Q96M61
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MAGEB18
- Synthetic peptide of Human MAGEB18
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice