Anti-TRIM27 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TRIM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-573
- 77848-572
- Antigen Symbol:
- TRIM27
- TRIM27
- Antigen Synonyms:
- AW538890|DAAP-182E11.3|MGC189472|OTTHUMP00000029314|Ret finger protein|RFP|RFP transforming protein|Rfp1|RING finger protein 76|RNF76|TRI27_HUMAN|TRIM 27|Tripartite motif containing 27|Tripartite motif containing protein 27|Tripartite motif protein Tripartite motif-containing protein 27|Zinc finger protein RFP
- AW538890|DAAP-182E11.3|MGC189472|OTTHUMP00000029314|Ret finger protein|RFP|RFP transforming protein|Rfp1|RING finger protein 76|RNF76|TRI27_HUMAN|TRIM 27|Tripartite motif containing 27|Tripartite motif containing protein 27|Tripartite motif protein Tripartite motif-containing protein 27|Zinc finger protein RFP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14373
- P14373
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TRIM27
- Synthetic peptide of Human TRIM27
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice