Anti-MRPS28 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPS28
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-377
- 77845-378
- Antigen Symbol:
- MRPS28
- MRPS28
- Antigen Synonyms:
- 28S ribosomal protein S28|28S ribosomal protein S35|HSPC007|Mitochondrial 28S ribosomal protein S35|mitochondrial|Mitochondrial ribosomal protein S28|MRP-S28|MRP-S35|MRPS 28|MRPS35|RT28_HUMAN|S28mt|S35mt
- 28S ribosomal protein S28|28S ribosomal protein S35|HSPC007|Mitochondrial 28S ribosomal protein S35|mitochondrial|Mitochondrial ribosomal protein S28|MRP-S28|MRP-S35|MRPS 28|MRPS35|RT28_HUMAN|S28mt|S35mt
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Q9
- Q9Y2Q9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 28S ribosomal protein S28, mitochondrial protein (72-187AA)
- Recombinant Human 28S ribosomal protein S28, mitochondrial protein (72-187AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice