Anti-FZD1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: FZD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-354
- 77844-353
- Antigen Symbol:
- FZD1
- FZD1
- Antigen Synonyms:
- Chfz-1|Dfrizzled-1|DKFZp564G072|FLJ95923|Frizzled 1|Frizzled homolog 1 (Drosophila)|Frizzled homolog 1|Frizzled-1|Fz receptor 1|Fz-1|Fz1|FZD1_HUMAN|FzE1|hFz1|Rfz1|Wnt receptor|Xfrizzled-1
- Chfz-1|Dfrizzled-1|DKFZp564G072|FLJ95923|Frizzled 1|Frizzled homolog 1 (Drosophila)|Frizzled homolog 1|Frizzled-1|Fz receptor 1|Fz-1|Fz1|FZD1_HUMAN|FzE1|hFz1|Rfz1|Wnt receptor|Xfrizzled-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UP38
- Q9UP38
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Frizzled-1 protein (73-322AA)
- Recombinant human Frizzled-1 protein (73-322AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice