Anti-MAGED2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: MAGED2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-668
- 77844-667
- Antigen Symbol:
- MAGED2
- MAGED2
- Antigen Synonyms:
- 11B6|BCG 1|BCG-1|Breast cancer-associated gene 1 protein|HCA10|Hepatocellular carcinoma-associated protein JCL-1|JCL 1|MAGD2_HUMAN|MAGE D|MAGE D2 antigen|MAGE-D2 antigen|MAGED2|Melanoma antigen family D 2|Melanoma associated antigen D2|Melanoma-associated antigen D2
- 11B6|BCG 1|BCG-1|Breast cancer-associated gene 1 protein|HCA10|Hepatocellular carcinoma-associated protein JCL-1|JCL 1|MAGD2_HUMAN|MAGE D|MAGE D2 antigen|MAGE-D2 antigen|MAGED2|Melanoma antigen family D 2|Melanoma associated antigen D2|Melanoma-associated antigen D2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNF1
- Q9UNF1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanoma-associated antigen D2 protein (38-285AA)
- Recombinant Human Melanoma-associated antigen D2 protein (38-285AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice