Anti-ATP5B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-602
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP5B
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P06576
- Antigen Synonyms:ATP 5B|ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide|ATP synthase subunit beta mitochondrial|ATP synthase subunit beta|mitochondrial|atp5b|ATPB|ATPB_HUMAN|ATPMB|ATPSB|Epididymis secretory protein Li 271|HEL-S-271|Mitochondrial ATP synthase beta subunit|Mitochondrial ATP Synthase Subunit Beta|Mitochondrial ATP synthetase beta subunit
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP synthase subunit beta, mitochondrial protein (230-529AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATP5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human