Anti-PAK6 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: PAK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77849-218
- 77849-217
- Antigen Symbol:
- PAK6
- PAK6
- Antigen Synonyms:
- CDKN1A activated kinase 6|EC 2.7.1.37|p21 activated protein kinase 6|p21 protein (Cdc42/Rac)-activated kinase 6|p21(CDKN1A) activated kinase 6|p21-activated kinase 6|p21activated kinase 6|PAK 5|PAK 6|PAK-5|PAK-6|PAK5|Pak6|PAK6_HUMAN|Serine threonine protein kinase PAK 6|Serine/threonine protein kinase PAK 6|Serine/threonine protein kinase PAK6|Serine/threonine-protein kinase PAK 6
- CDKN1A activated kinase 6|EC 2.7.1.37|p21 activated protein kinase 6|p21 protein (Cdc42/Rac)-activated kinase 6|p21(CDKN1A) activated kinase 6|p21-activated kinase 6|p21activated kinase 6|PAK 5|PAK 6|PAK-5|PAK-6|PAK5|Pak6|PAK6_HUMAN|Serine threonine protein kinase PAK 6|Serine/threonine protein kinase PAK 6|Serine/threonine protein kinase PAK6|Serine/threonine-protein kinase PAK 6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQU5
- Q9NQU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PAK6
- Fusion protein of Human PAK6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice