Anti-SLC6A13 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC6A13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-483
- 77843-484
- Antigen Symbol:
- SLC6A13
- SLC6A13
- Antigen Synonyms:
- GABA transport protein|GAT 2|GAT-2|GAT2|S6A13_HUMAN|Sodium- and chloride-dependent GABA transporter 2|Solute carrier family 6 (Neurotransmitter transporter|GABA)|member 13|Solute carrier family 6 member 13
- GABA transport protein|GAT 2|GAT-2|GAT2|S6A13_HUMAN|Sodium- and chloride-dependent GABA transporter 2|Solute carrier family 6 (Neurotransmitter transporter|GABA)|member 13|Solute carrier family 6 member 13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NSD5
- Q9NSD5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sodium- and chloride-dependent GABA transporter 2 protein (1-40AA)
- Recombinant Human Sodium- and chloride-dependent GABA transporter 2 protein (1-40AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice