Anti-GLP1R Rabbit Recombinant Polyclonal Antibody
Catalog # 77856-093
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GLP1R
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P43220
- Antigen Synonyms:GLP 1 R|GLP 1 receptor|GLP 1R|GLP|GLP-1 receptor|GLP-1-R|GLP-1R|GLP1R_HUMAN|Glucagon like peptide 1 receptor|Glucagon-like peptide 1 receptor|MGC138331|OTTHUMP00000016340
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Glucagon-like peptide 1 receptor protein (24-145AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: GLP1R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human