Anti-PPIE Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPIE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-137
- 77844-136
- Antigen Symbol:
- PPIE
- PPIE
- Antigen Synonyms:
- Ab1-210|Cyclophilin 33|Cyclophilin E|Cyclophilin-33|CYP 33|CYP33|EC 5.2.1.8|Peptidyl prolyl cis trans isomerase E|Peptidyl-prolyl cis-trans isomerase E|Peptidylprolyl isomerase E (cyclophilin E)|Peptidylprolyl isomerase E|PPIase E|PPIE|PPIE_HUMAN|Rotamase E
- Ab1-210|Cyclophilin 33|Cyclophilin E|Cyclophilin-33|CYP 33|CYP33|EC 5.2.1.8|Peptidyl prolyl cis trans isomerase E|Peptidyl-prolyl cis-trans isomerase E|Peptidylprolyl isomerase E (cyclophilin E)|Peptidylprolyl isomerase E|PPIase E|PPIE|PPIE_HUMAN|Rotamase E
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNP9
- Q9UNP9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Peptidyl-prolyl cis-trans isomerase E protein (86-141AA)
- Recombinant Human Peptidyl-prolyl cis-trans isomerase E protein (86-141AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice