Anti-ANXA9 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-196
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ANXA9
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O76027
- Antigen Synonyms:annexin 31|formerly|Annexin A9|Annexin XXXI|Annexin-31|Annexin-9|ANX31|ANXA9_HUMAN|Pemphaxin
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Annexin A9 protein (8-345AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: ANXA9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse