Anti-BACE2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77845-730
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BACE2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y5Z0
- Antigen Synonyms:ALP 56|ALP56|Asp 1|ASP1|ASP21|Aspartic Like Protease 56kDa|Aspartic-like protease 56 kDa|Aspartyl protease 1|BACE 2|BACE2_HUMAN|Beta Secretase 2|Beta Site Amyloid Beta A4 Precursor Protein Cleaving Enzyme 2|Beta-secretase 2|Beta-site amyloid precursor protein cleaving enzyme 2|Beta-site APP cleaving enzyme 2|Down region aspartic protease|Downs Syndrome Region Aspartic Protease|DRAP|Memapsin 1|Memapsin-1|Membrane associated aspartic protease 1|Membrane-associated aspartic protease 1|Theta-secretase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-secretase 2 protein (84-157AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: BACE2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat